Anti-TICAM-2 Mouse Monoclonal Antibody [clone: 9G1B30]

Supplier: BioLegend
659402 659402
10763-778EA 285.98 USD
CA10763-778 10763-778
Anti-TICAM-2 Mouse Monoclonal Antibody [clone: 9G1B30]
Antibodies
Purified anti-TICAM-2 (TRAM) [9G1B30]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TICAM-2
Clonality: Monoclonal
Clone: 9G1B30
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR