Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGF-beta1 Mouse Monoclonal Antibody () [clone: 21C11]

Supplier: Biolegend
525301
Purified anti-TGF-β1 [21C11]; Isotype: Mouse IgG2a, κ; Reactivity: Mammalian species; Apps: ELISA Capture; Size: 50 μg

Each lot of this antibody is quality control tested by ELISA assay. A concentration of 1 μg/ml of the capture antibody was utilized to generate the example standard curve. It is recommended that the end-user titrate each lot of reagents for optimal performance.

Type: Primary
Antigen: TGF-beta1
Clonality: Monoclonal
Clone: 21C11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mammalian species




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 μg 525301 10761-776 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TGF-beta1
Antigen Name TGF-beta1
Reactivity Mammalian species
Clonality Monoclonal
Antibody Type Primary
Clone 21C11
Host Mouse
Isotype Mouse IgG2a, κ
ELISA Yes
Gel Shift Yes
Antigen Synonyms Transforming growth factor-beta 1 (TGF-b1), TGFB1, CED, DPD1
Gene ID 7040, 21803
Concentration 0.50 mg/mL
Immunogen Human TGF-β1
Purification The antibody was purified by antigen-affinity chromatography.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty