Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGF-β Mouse Monoclonal Antibody (LEAF) [clone: TW7-7H4]

Supplier: Biolegend
361202 361201
LEAF™ Purified anti-human/mouse Latent TGF-β [TW7-7H4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay.  Concentrations of 1 μg/ml and 0.5 μg/ml of the capture antibody were utilized to generate the example standard curves for human latent TGF-β and mouse latent TGF-β, respectively. It is recommended that each lot of reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TGF-β
Clonality: Monoclonal
Clone: TW7-7H4
Conjugation: LEAF
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 μg 361202 10760-842 Each (500µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TGF-β
Antigen Name TGF-β
Reactivity Mouse, Human
Clonality Monoclonal
Antibody Type Primary
Clone TW7-7H4
Host Mouse
Isotype Mouse IgG1, κ
ELISA Yes
Gel Shift Yes
Antigen Synonyms Latent TGF-β1, Latent TGFβ, Latent TGFβ1, LTGF-β1, LTGF-β, LTGFβ1, LTGFβ, Latent TGF-b1, Latent TGF-b, Latent TGFb1, Latent TGFb, LTGF-b1, LTGF-b, LTGFb1, LTGFb, Latent TGF-beta, Latent TGF-beta1, Latent TGFbeta, Latent TGFbeta1, LTGF-beta1, LTGFbeta1, LTGFbeta, Latent transforming growth factor beta
Gene ID 7040
Concentration 1.00 mg/mL
Immunogen Mouse Latent TGF-β
Purification The LEAF™ (Low Endotoxin, Azide-Free) antibody was purified by affinity chromatography.
Form 0.2 µm filtered in phosphate-buffered solution, pH 7.2, containing no preservative. Endotoxin level is <0.1 EU/µg of the protein (<0.01 ng/µg of the protein) as determined by the LAL test.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C. This LEAF™ solution contains no preservative; handle under aseptic conditions.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping