|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | TCR-b chain |
Antigen Name | TCR-b chain |
Reactivity | Mouse |
Conjugation | Alexa Fluor® 594 |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | H57-597 |
Host | Armenian Hamster |
Isotype | Armenian Hamster IgG |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Flow Cytometry | Yes |
Antigen Synonyms | TCR-β chain, TCR-β, β-TCR |
Gene ID | 21577 |
Concentration | 0.50 mg/mL |
Immunogen | Affinity purified TCR from mouse DO-11.10 cells |
Purification | The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions. The solution is free of unconjugated Alexa Fluor® 594. |
Form | Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. |
Storage Temperature | The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. <b>Do not freeze.</b> |
Shipping Temperature | Ambient RT |