Anti-TCR Vg1.1 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2.11]

Supplier: BioLegend
141104 141103
141103-BLEA 120.94 USD
141103-BL 141104-BL
Anti-TCR Vg1.1 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2.11]
Antibodies
FITC anti-mouse TCR Vγ1.1/Cr4 [2.11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR Vg1.1
Clonality: Monoclonal
Clone: 2.11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR