Anti-TCR Vb12 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MR11-1]

Supplier: BioLegend
139703 139704 139704 139703
139703-BLEA 139.18 USD
CA139704-BL CA139703-BL 139703-BL 139704-BL
Anti-TCR Vb12 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MR11-1]
Antibodies
PE anti-mouse TCR Vβ12 [MR11-1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR Vb12
Clonality: Monoclonal
Clone: MR11-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR