Purified anti-T-bet (MaxPar® Ready) [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, CyTOF®; Size: 100 μg
This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.
Type: Primary
Antigen: T-bet
Clonality: Monoclonal
Clone: 4B10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse