Anti-T-bet Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 4B10]

Supplier: BioLegend
644819 644819
644819-BLEA 344.1 USD
644819-BL CA644819-BL
Anti-T-bet Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: 4B10]
Antibodies
Brilliant Violet 711™ anti-T-bet [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 125 μl

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our our <a href="http://www.biolegend.com/media_assets/support_protocol/True-Nuclear_Trademark_Transcription_Factor_Staining_Protocol.pdf" target="_blank">True-Nuclear™ Transcription Factor Staining Protocol</a>. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: T-bet
Clonality: Monoclonal
Clone: 4B10
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR