Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SWAP70 Mouse Monoclonal Antibody [clone: 9D9A19]

Supplier: Biolegend
654702 654702
10763-512 CA10763-512
Anti-SWAP70 Mouse Monoclonal Antibody [clone: 9D9A19]
Purified anti-SWAP70 [9D9A19]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1-0.5 μg per ml. For immunoprecipitation, the suggested use of this reagent is 10-30 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SWAP70
Clonality: Monoclonal
Clone: 9D9A19
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty