Anti-SUSD2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: W5C5]

Supplier: BioLegend
327408 327408
10758-344EA 377.86 USD
10758-344 CA10758-344
Anti-SUSD2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: W5C5]
Antibodies
APC anti-human SUSD2 [W5C5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SUSD2
Clonality: Monoclonal
Clone: W5C5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR