Anti-STAT6 Mouse Monoclonal Antibody [clone: 16G12A08]

Supplier: BioLegend
657902 657902
10763-764EA 302.62 USD
CA10763-764 10763-764
Anti-STAT6 Mouse Monoclonal Antibody [clone: 16G12A08]
Antibodies
Purified anti-STAT6 [16G12A08]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. For immunoprecipitation, the suggested use of this reagent is 5-20 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: STAT6
Clonality: Monoclonal
Clone: 16G12A08
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR