Anti-STAT4 Mouse Monoclonal Antibody [clone: 15A1B41]

Supplier: BioLegend
660502 660502
10763-354EA 258.15 USD
10763-354 CA10763-354
Anti-STAT4 Mouse Monoclonal Antibody [clone: 15A1B41]
Purified anti-STAT4 [15A1B41]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1 - 0.5 μg per ml. For immunofluorescent staining, the suggested use of this reagent is 1-5 μg per ml. For immunoprecipitation, the suggested use of this reagent is 10-30 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: STAT4
Clonality: Monoclonal
Clone: 15A1B41
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR