Anti-STAT1 Mouse Monoclonal Antibody [clone: 10C4B40]

Supplier: BioLegend
661002 661001 661002 661001
10763-614EA 285.98 USD
CA10763-614 CA10763-612 10763-614 10763-612
Anti-STAT1 Mouse Monoclonal Antibody [clone: 10C4B40]
Antibodies
Purified anti-STAT1 [10C4B40]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 1.0 μg per ml (1:500). It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 10C4B40
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR