Anti-SSRP1 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 3E4]

Supplier: BioLegend
609806 609806
10762-770EA 338.48 USD
10762-770 CA10762-770
Anti-SSRP1 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 3E4]
Alexa Fluor® 594 anti-SSRP1 [3E4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SSRP1
Clonality: Monoclonal
Clone: 3E4
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR