Anti-SSEA5 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 8E11]

Supplier: BioLegend
355205 355206 355205 355206
10759-786EA 159.4 USD
CA10759-786 10759-786 10759-788 CA10759-788
Anti-SSEA5 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 8E11]
Type: Primary
Antigen: SSEA5
Clonality: Monoclonal
Clone: 8E11
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR