Anti-SSEA-5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 8E11]

Supplier: BioLegend
355204 355203 355204 355203
10759-598EA 165.11 USD
10759-598 10759-784 CA10759-784 CA10759-598
Anti-SSEA-5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 8E11]
Antibodies
PE anti-human SSEA-5 [8E11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SSEA-5
Clonality: Monoclonal
Clone: 8E11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR