Anti-SPHK1 Mouse Monoclonal Antibody [clone: 1A5SC]

Supplier: BioLegend
658302 658302
10763-586EA 335.79 USD
10763-586 CA10763-586
Anti-SPHK1 Mouse Monoclonal Antibody [clone: 1A5SC]
Antibodies
Purified anti-SPHK1 [1A5SC]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SPHK1
Clonality: Monoclonal
Clone: 1A5SC
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR