Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX2 Mouse Monoclonal Antibody (Alexa Fluor® 594) [clone: 14A6A34]

Supplier: Biolegend
656106
Alexa Fluor® 594 anti-SOX2 [14A6A34]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: 14A6A34
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 656106 10763-756 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SOX2
Antigen Name SOX2
Reactivity Human, Mouse
Conjugation Alexa Fluor® 594
Clonality Monoclonal
Antibody Type Primary
Clone 14A6A34
Host Mouse
Isotype Mouse IgG1, κ
ImmunoFluorescence Yes
Antigen Synonyms SRY-related HMG-box gene 2, SRY (sex determining region Y)-box 2, MCOPS3, ANOP3
Gene ID 6657
Concentration 0.50 mg/mL
Immunogen Full length human SOX recombinant protein
Purification The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions. The solution is free of unconjugated Alexa Fluor® 594.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. <b>Do not freeze.</b>
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty