Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOX2 Rabbit Polyclonal Antibody () [clone: Poly6519]

Supplier: Biolegend
651902 651901
Purified anti-SOX2 [Poly6519]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human; Apps: WB, IF; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescent staining, the suggested use of this reagent is 1 - 5 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SOX2
Clonality: Polyclonal
Clone: Poly6519
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
200 μl 651902 651902-BL Each (200µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

50 μl 651901 651901-BL Each (50µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SOX2
Antigen Name SOX2
Reactivity Human
Clonality Polyclonal
Antibody Type Primary
Clone Poly6519
Host Rabbit
Isotype Rabbit Polyclonal IgG
Western Blot Yes
ImmunoFluorescence Yes
Antigen Synonyms SRY (sex determining region Y)-box 2, ANOP3, SOX-2
Gene ID 6657
Concentration 0.00 mg/mL
Immunogen Full length recombinant SOX2 protein
Form This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Storage Temperature Upon receipt, store undiluted at -20°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping