Anti-Siglec-10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5G6]

Supplier: BioLegend
347606 347606
347606-BLEA 396.31 USD
347606-BL CA347606-BL
Anti-Siglec-10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5G6]
Antibodies
APC anti-human Siglec-10 [5G6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Siglec-10
Clonality: Monoclonal
Clone: 5G6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR