|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | Siglec H | |
Antigen Name | Siglec H | |
Reactivity | Mouse | |
Conjugation | APC (Allophycocyanin) | |
Clonality | Monoclonal | |
Antibody Type | Primary | |
Clone | 551 | |
Host | Rat | |
Isotype | Rat IgG1 | |
Antigen Synonyms | Siglec-H, Sialic acid-binding immunoglobulin-like lectin | |
Gene ID | 233274 | |
Concentration | 0.20 mg/mL | |
Immunogen | Siglec-H-Fc recombinant fusion protein | |
Purification | The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody. | |
Form | Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. | |
Storage Temperature | The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. <B>Do not freeze.</B> | |
Shipping Temperature | Ambient RT |