Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-INPP5D Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1C1-A5]

Supplier: Biolegend
Type: Primary
Antigen: INPP5D
Clonality: Monoclonal
Clone: P1C1-A5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 656604 10763-550 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 656603 10763-548 Each (25µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol INPP5D
Antigen Name Inositol Polyphosphate-5-Phosphatase D
Reactivity Human, Mouse Mouse, Human
Conjugation PE (Phycoerythrin)
Clonality Monoclonal
Antibody Type Primary
Clone P1C1-A5
Host Mouse
Isotype IgG1 kappa
Antigen Synonyms p150Ship, SHIP-1, SH2 domain-containing inositol-5'-phosphatase 1, INPP5D, Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 1, SH2 domain-containing inositol phosphatase 1, SHIP1, hp51CN, Inositol polyphosphate-5-phosphatase of 145 kDa, SHIP, SIP-145 SHIP1, SH2 domain-containing inositol phosphatase 1, hp51CN, SHIP, p150Ship, SIP-145, INPP5D, Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 1, SHIP-1, SH2 domain-containing inositol-5'-phosphatase 1, Inositol polyphosphate-5-phosphatase of 145 kDa
Gene ID 3635
Concentration 0.2 mg/ml
Immunogen Human SHIP fusion protein in CFA/IFA
Purification The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty