Anti-RUNX1 Mouse Monoclonal Antibody [clone: 1C5B16]

Supplier: BioLegend
659302 659302
10763-776EA 285.98 USD
CA10763-776 10763-776
Anti-RUNX1 Mouse Monoclonal Antibody [clone: 1C5B16]
Antibodies
Purified anti-RUNX1 [1C5B16]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Mouse (weak); Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.25 - 1.0 μg per ml. For immunoprecipitation, the suggested use of this reagent is 10-30 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: RUNX1
Clonality: Monoclonal
Clone: 1C5B16
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Mouse (weak)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR