Anti-RORA Rabbit Polyclonal Antibody [clone: Poly6388]

Supplier: BioLegend
638802 638801
CA638801-BL CA638802-BL
Anti-RORA Rabbit Polyclonal Antibody [clone: Poly6388]
Type: Primary
Antigen: RORA
Clonality: Polyclonal
Clone: Poly6388
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR