|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PSMA | |
Antigen Name | PSMA | |
Reactivity | Human | |
Conjugation | PE (Phycoerythrin)/Cy7® | |
Clonality | Monoclonal | |
Antibody Type | Primary | |
Clone | LNI-17 | |
Host | Mouse | |
Isotype | Mouse IgG1, κ | |
Antigen Synonyms | Prostate-specific membrane antigen, PSM, FOLH1, Folate hydrolase 1, GCP2, Glutamate carboxypeptidase 2, NAALADase I, FGCP | |
Gene ID | 2346 | |
Concentration | 0.00 μg/mL | |
Purification | The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody. | |
Form | Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). | |
Storage Temperature | The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. <b>Do not freeze.</b> | |
Shipping Temperature | Ambient RT |