Anti-PLCG1 Rabbit Polyclonal Antibody [clone: Poly6562]

Supplier: BioLegend
656202 656202
10763-636EA 371.95 USD
CA10763-636 10763-636
Anti-PLCG1 Rabbit Polyclonal Antibody [clone: Poly6562]
Type: Primary
Antigen: PLCG1
Clonality: Polyclonal
Clone: Poly6562
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR