Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PERK Phospho(Ser713) Rabbit Polyclonal Antibody () [clone: Poly6494]

Supplier: Biolegend
649402 649401
Purified anti-PERK Phospho (Ser713) [Poly6494]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human; Apps: WB; Size: 400 μl

Each lot of this antibody is quality control tested by <a title="View Protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_Western_blotting_protocol.pdf" target="_blank">Western blotting</a>. Western blotting, suggested working dilution(s): Use 20 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: PERK Phospho(Ser713)
Clonality: Polyclonal
Clone: Poly6494
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl 649401 649401-BL Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

400 μl 649402 649402-BL Each (400µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PERK Phospho(Ser713)
Antigen Name PERK Phospho(Ser713)
Reactivity Human
Clonality Polyclonal
Antibody Type Primary
Clone Poly6494
Host Rabbit
Isotype Rabbit Polyclonal IgG
Western Blot Yes
Antigen Synonyms eukaryotic translation initiation factor 2 alpha kinase 3, heme-regulated EIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase PRKR-like endoplasmic reticulum kinase, eukaryotic translation initiation factor 2 alpha kinase 3, heme-regulated EIF2-alpha kinase
Gene ID 9451
Concentration 0.00 mg/mL
Immunogen Modified synthetic peptide
Purification The antibody was purified by affinity chromatography.
Form This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Storage Temperature Upon receipt, store at -20°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty