Anti-PD-1H Armenian Hamster Monoclonal Antibody (LEAF) [clone: MH5A]

Supplier: BioLegend
143704 143703 143704 143703
143704-BLEA 605.13 USD
CA143703-BL CA143704-BL 143704-BL 143703-BL
Anti-PD-1H Armenian Hamster Monoclonal Antibody (LEAF) [clone: MH5A]
LEAF™ Purified anti-mouse PD-1H (VISTA) [MH5A]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC, WB, Block; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: PD-1H
Clonality: Monoclonal
Clone: MH5A
Conjugation: LEAF
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR