Purified anti-mouse CD279 (PD-1) (MaxPar® Ready) [RMP1-30]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, CyTOF®; Size: 100 μg
This product is suitable for use with the <a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the <a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.
Type: Primary
Antigen: PD-1 (CD279)
Clonality: Monoclonal
Clone: RMP1-30
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse