Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PD1 Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: 29F.1A12]

Supplier: Biolegend
Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: 29F.1A12
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 135224 10756-422 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 135223 10756-420 Each (25µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PD1
Antigen Name Programmed cell death protein 1
Reactivity Mouse
Conjugation APC (Allophycocyanin)/Cy7®
Clonality Monoclonal
Antibody Type Primary
Clone 29F.1A12
Host Rat
Isotype IgG2a kappa
Antigen Synonyms CD279, hPD-1, protein PD-1, PD-1, hPD-l, programmed cell death protein 1, SLEB2, PD1 SLEB2, hPD-1, protein PD-1, programmed cell death protein 1, PD1, CD279, PD-1, hPD-l
Gene ID 18566
Concentration 0.2 mg/ml
Immunogen PD-1 cDNA followed by PD-1-Ig fusion protein
Purification The antibody was purified by affinity chromatography and conjugated with APC/Cy7 under optimal conditions. The solution is free of unconjugated APC/Cy7 and unconjugated antibody.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty