|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | CD279 | |
Antigen Name | CD279 | |
Reactivity | Mouse | |
Conjugation | PE (Phycoerythrin)/Cy7® | |
Clonality | Monoclonal | |
Antibody Type | Primary | |
Clone | 29F.1A12 | |
Host | Rat | |
Isotype | Rat IgG2a, κ | |
Antigen Synonyms | Programmed Death-1, PD1 | |
Gene ID | 18566 | |
Concentration | 0.20 mg/mL | |
Immunogen | PD-1 cDNA followed by PD-1-Ig fusion protein | |
Purification | The antibody was purified by affinity chromatography, and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody. | |
Form | Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide. | |
Storage Temperature | The antibody solution should be stored undiluted between 2°C and 8°C, and protected from prolonged exposure to light. <B>Do not freeze.</b> | |
Shipping Temperature | Ambient RT |