Anti-PAX9 Mouse Monoclonal Antibody [clone: 14D10A24]

Supplier: BioLegend
658202 658202
10763-584EA 335.79 USD
CA10763-584 10763-584
Anti-PAX9 Mouse Monoclonal Antibody [clone: 14D10A24]
Antibodies
Purified anti-PAX9 [14D10A24]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: WB, IF; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. <span>For immunofluorescence microscopy, the suggested usage is 1 - 5 μg per mL. </span>It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: PAX9
Clonality: Monoclonal
Clone: 14D10A24
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR