Purified anti-Pax-5 [1H9]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, IF, IHC; Size: 100 μg
Each lot of this antibody is quality control tested by <a style="color: #4c0073; text-decoration: underline;" title="View Protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_Western_blotting_protocol.pdf" target="_blank">Western blotting</a>. Western blotting, suggested working dilution(s): Use 2-4 μl per ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Pax-5
Clonality: Monoclonal
Clone: 1H9
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Mouse