Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAR1 Mouse Monoclonal Antibody () [clone: N2-11]

Supplier: Biolegend
Purified anti-PAR1 [N2-11]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, IF, IHC, ELISA; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: PAR1
Clonality: Monoclonal
Clone: N2-11
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 659102 10763-770 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol PAR1
Antigen Name PAR1
Reactivity Human
Clonality Monoclonal
Antibody Type Primary
Clone N2-11
Host Mouse
Isotype Mouse IgG1
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Gel Shift Yes
Antigen Synonyms Protease-Activated Receptor 1, Coagulation Factor II (Thrombin) Receptor, Thrombin Receptor, F2R, CF2R
Gene ID 2149
Concentration 0.50 mg/mL
Immunogen Synthetic peptide conjugagted to KLH
Purification The antibody was purified by affinity chromatography.
Form This antibody is provided in phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Storage Temperature The antibody solution should be stored undiluted between 2°C and 8°C.
Shipping Temperature Ambient RT
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty