Anti-FBXO8 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP18733C
89515-742EA 636.79 USD
89515-742
Anti-FBXO8 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: FBXO8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR