Anti-Nucleolin Phospho (Thr76/Thr84) Mouse Monoclonal Antibody [clone: 10C7]

Supplier: BioLegend
609403 609404 609404 609403
609403-BLEA 151.75 USD
CA609403-BL CA609404-BL 609403-BL 609404-BL
Anti-Nucleolin Phospho (Thr76/Thr84) Mouse Monoclonal Antibody [clone: 10C7]
Purified anti-Nucleolin-Phospho (Thr76/Thr84) [10C7]; Isotype: Mouse IgM, κ; Reactivity: Human, reacts against Thr76/Thr84-phosphorylated nucleolin; Apps: WB, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For IHC, use 10 μg/ml of antibody for staining. Antigen retrieval using 0.01 M sodium citrate buffer is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Nucleolin Phospho (Thr76/Thr84)
Clonality: Monoclonal
Clone: 10C7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human;reacts against Thr76/Thr84-phosphorylated nucleolin
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR