Anti-MRGX2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: K125H4]

Supplier: BioLegend
359003 359004 359003 359004
10760-504EA 434.22 USD
CA10760-502 CA10760-504 10760-504 10760-502
Anti-MRGX2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: K125H4]
Antibodies
PE anti-human MRGX2 [K125H4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MRGX2
Clonality: Monoclonal
Clone: K125H4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR