Anti-LOX-1 Mouse Monoclonal Antibody [clone: 15C4]

Supplier: BioLegend
358602 358602
10760-444EA 285.98 USD
CA10760-444 10760-444
Anti-LOX-1 Mouse Monoclonal Antibody [clone: 15C4]
Antibodies
Purified anti-human LOX-1 [15C4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: LOX-1
Clonality: Monoclonal
Clone: 15C4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR