Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD39 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: A1]

Supplier: Biolegend

328219 328219
10758-364EA 212.41 USD InStock
10758-364 CA10758-364
Anti-CD39 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: A1]
Antibodies
Brilliant Violet 510™ anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rhesus; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD39
Clonality: Monoclonal
Clone: A1
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Rhesus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty