Anti-CD38 Mouse Monoclonal Antibody [clone: 14.27]

Supplier: BioLegend
250502 250502
10757-550EA 209.51 USD
10757-550 CA10757-550
Anti-CD38 Mouse Monoclonal Antibody [clone: 14.27]
Antibodies
Purified anti-rat CD38 [14.27]; Isotype: Mouse IgG2b, κ; Reactivity: Rat; Apps: FC, IHC, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 14.27
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR