Anti-CD371 (CLEC12A) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 50C1]

Supplier: BioLegend
353609 353610 353610 353609
10759-702EA 171.67 USD
CA10759-702 10759-702 10759-704 CA10759-704
Anti-CD371 (CLEC12A) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 50C1]
Antibodies
PE/Cy7 anti-human CD371 (CLEC12A) [50C1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD371 (CLEC12A)
Clonality: Monoclonal
Clone: 50C1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR