Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 449CT13.4.2]

Supplier: Abcepta
AM2018A
89512-178EA 606.77 USD
89512-178
Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 449CT13.4.2]
Antibodies
Western Blot: 1:500~4000,

Type: Primary
Antigen: AKT1S1
Clonality: Monoclonal
Clone: 449CT13.4.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR