Anti-CIDEC Rabbit Polyclonal Antibody

Supplier: Abcepta
AP18249C
89516-984EA 636.79 USD
89516-984
Anti-CIDEC Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR