Anti-CD369 (Dectin-1, CLEC7A) Rat Monoclonal Antibody [clone: RH1]

Supplier: BioLegend
144302 144302 144301 144301
10759-342EA 235.31 USD
CA10759-342 10759-342 CA10759-340 10759-340
Anti-CD369 (Dectin-1, CLEC7A) Rat Monoclonal Antibody [clone: RH1]
Antibodies
Purified anti-mouse CD369 (Dectin-1, CLEC7A) [RH1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD369 (Dectin-1, CLEC7A)
Clonality: Monoclonal
Clone: RH1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR