Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD368 (CLEC4D) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9B9]

Supplier: Biolegend

360206 360205 360205 360206
CA10760-722 10760-720 CA10760-720 10760-722
Anti-CD368 (CLEC4D) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9B9]
Antibodies
APC anti-human CD368 (CLEC4D) [9B9]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD368 (CLEC4D)
Clonality: Monoclonal
Clone: 9B9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty