Anti-CD366 (Tim-3) Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: F38-2E2]

Supplier: BioLegend
345016 345016 345015 345015
10758-756EA 171.67 USD
10758-756 10758-758 CA10758-756 CA10758-758
Anti-CD366 (Tim-3) Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: F38-2E2]
Antibodies
PerCP/Cy5.5 anti-human CD366 (Tim-3) [F38-2E2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD366 (Tim-3)
Clonality: Monoclonal
Clone: F38-2E2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR