Anti-RNF166 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20484C
10217-672EA 577.43 USD
10217-672
Anti-RNF166 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunofluorescence: 1:25,

Type: Primary
Antigen: RNF166
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR