Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD360 Mouse Monoclonal Antibody (LEAF) [clone: 17A12]

Supplier: Biolegend

359503 359504 359503 359504
10760-606 10760-608 CA10760-606 CA10760-608
Anti-CD360 Mouse Monoclonal Antibody (LEAF) [clone: 17A12]
LEAF™ Purified anti-human CD360 (IL-21R) [17A12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IF; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD360
Clonality: Monoclonal
Clone: 17A12
Conjugation: LEAF
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty