Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD36 Mouse Monoclonal Antibody [clone: 5-271]

Supplier: Biolegend

336215 336215
CA10758-548 10758-548
Anti-CD36 Mouse Monoclonal Antibody [clone: 5-271]
Purified anti-human CD36 (MaxPar® Ready) [5-271]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, CyTOF®; Size: 100 μg

<span>This product is suitable for use with the </span><a href="http://www.dvssciences.com/labeling-reagents.php" target="_blank">MaxPar® Metal Labeling Kits</a><span>. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the </span><a href="http://www.dvssciences.com/technical-literature_13_3283624161.pdf" target="_blank">MaxPar® antibody labeling protocol</a><span>). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.</span>

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 5-271
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty