Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD339 Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: HMJ1-29]

Supplier: Biolegend

130914 130914 130913 130913
10757-166EA 333.28 USD InStock
10757-166 CA10757-166 10757-164 CA10757-164
Anti-CD339 Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: HMJ1-29]
Antibodies
APC anti-mouse CD339 (Jagged 1) [HMJ1-29]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD339
Clonality: Monoclonal
Clone: HMJ1-29
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty